Learn More
Cinacalcet Hydrochloride, Mikromol™
Description
Cinacalcet Hydrochloride
Specifications
Specifications
| Chemical Name or Material | Cinacalcet Hydrochloride |
| CAS | 364782-34-3 |
| Analyte or Component Names | Cinacalcet Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C22 H22 F3 N . Cl H |
| Certifications/Compliance | ISO 17034 |
| Synonym | AMG 073 HCl, Cinacalcet hydrochloride, KRN 1493, Mimpara, N-[(1R)-1-(1-Naphthyl)ethyl]-3-[3-(trifluoromethyl)phenyl]propan-1-amine hydrochloride, Regpara, Sensipar, 1-Naphthalenemethanamine, alpha-methyl-N-[3-[3-(trifluoromethyl)phenyl]propyl]-, hydrochloride (1:1), (alphaR)-, 1-Naphthalenemethanamine, alpha-methyl-N-[3-[3-(trifluoromethyl)phenyl]propyl]-, hydrochloride, (alphaR)- (9CI) |
| InChI Formula | InChI=1S/C22H22F3N.ClH/c1-16(20-13-5-10-18-9-2-3-12-21(18)20)26-14-6-8-17-7-4-11-19(15-17)22(23,24)25;/h2-5,7,9-13,15-16,26H,6,8,14H2,1H3;1H/t16-;/m1./s1 |
| SMILES | Cl.C[C@@H](NCCCc1cccc(c1)C(F)(F)F)c2cccc3ccccc23 |
| IUPAC Name | N-[(1R)-1-naphthalen-1-ylethyl]-3-[3-(trifluoromethyl)phenyl]propan-1-amine;hydrochloride |
| Show More |