Learn More
Cinanserin, TRC
Product Code. 30356962
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30356962 25 mg 25mg
30350460 100 mg -
30353993 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30356962 Supplier Toronto Research Chemicals Supplier No. TRCC44190325MG
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1166-34-3
Molecular Formula C20H24N2OS
Molecular Weight (g/mol) 340.48
Synonym N-[2-[[3-(Dimethylamino)propyl]thio]phenyl]-3-phenyl-2-propenamide, 2'-[[3-(Dimethylamino)propyl]thio]cinnamanilide, Cinanserin, Cinanserine, 2'-[[3-(Dimethylamino)propyl]thio]-cinnamanilide
IUPAC Name (E)-N-[2-[3-(dimethylamino)propylsulfanyl]phenyl]-3-phenylprop-2-enamide
SMILES CN(C)CCCSc1ccccc1NC(=O)\C=C\c2ccccc2
Specifications
Quantity 25 mg
InChI Formula InChI=1S/C20H24N2OS/c1-22(2)15-8-16-24-19-12-7-6-11-18(19)21-20(23)14-13-17-9-4-3-5-10-17/h3-7,9-14H,8,15-16H2,1-2H3,(H,21,23)/b14-13+
Formula Weight 340.1609
Chemical Name or Material Cinanserin