missing translation for 'onlineSavingsMsg'
Learn More

Cinnamyl acetate, 95%, BLD Pharm™

Product Code. 31032966
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
Unit Size:
100g
1kg
25g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31032966 500 g -
31028986 100 g 100g
31032026 1 kg 1kg
31030278 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 31032966 Supplier BLD Pharm Supplier No. BD121365500g
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 103-54-8
Molecular Formula C11H12O2
MDL Number MFCD00008722
Quantity 500 g
SMILES CC(OC/C=C/C1=CC=CC=C1)=O
Molecular Weight (g/mol) 176.21
Percent Purity 95%
Chemical Name or Material Cinnamyl acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.