missing translation for 'onlineSavingsMsg'
Learn More

Cinnarizine EP Impurity E, Mikromol™

Product Code. 30049880
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30049880 25 mg 25mg
17992066 100 mg 100mg
30462187 4 x 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30049880 Supplier Mikromol Supplier No. MM1038.060025
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Cinnarizine EP Impurity E
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,4-Bis(diphenylmethyl)piperazine
CAS 216581-01-0
Analyte or Component Names 1,4-Bis(diphenylmethyl)piperazine
Shipping Condition Room Temperature
Molecular Formula C30 H30 N2
Certifications/Compliance ISO 17025
Synonym 1,4-Bis(diphenylmethyl)piperazine, Cinnarizine Imp. E (EP), Cinnarizine Impurity E
InChI Formula InChI=1S/C30H30N2/c1-5-13-25(14-6-1)29(26-15-7-2-8-16-26)31-21-23-32(24-22-31)30(27-17-9-3-10-18-27)28-19-11-4-12-20-28/h1-20,29-30H,21-24H2
SMILES C1CN(CCN1C(c2ccccc2)c3ccccc3)C(c4ccccc4)c5ccccc5
IUPAC Name 1,4-dibenzhydrylpiperazine
Molecular Weight (g/mol) 418.57
Quantity 25 mg
Formula Weight 418.2409 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.