missing translation for 'onlineSavingsMsg'
Learn More

cis-1,4-Dichloro-2-butene, 95%, AcroSeal™, ACROS Organics™

Product Code. 10319847 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
100mL
Product Code. Quantity Packaging unitSize
10319847 100 mL AcroSeal™ Glass Bottle 100mL
1 options
This item is not returnable. View return policy

Product Code. 10319847

Brand: Thermo Scientific Acros 430741000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 1476-11-5
Color Colorless to Yellow
Density 1.18g/mL
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging AcroSeal™ Glass Bottle
Molecular Formula C4H6Cl2
Linear Formula ClCH2CH=CHCH2Cl
Refractive Index 1.4884 to 1.4914 (20°C, 589nm)
Quantity 100 mL
Synonym cis-1,4-dichloro-2-butene, z-1,4-dichlorobut-2-ene, 1,4-dichloro-cis-2-butene, 2-butene, 1,4-dichloro-, z, cis-1,4-dichlorobut-2-ene, unii-bqs2752i90, ccris 2651, cis 1,4-dichlorobutene, 2-butene, 1,4-dichloro-, cis, cis-2,3-dichlorobut-2-ene
InChI Key FQDIANVAWVHZIR-UPHRSURJSA-N
SMILES C(C=CCCl)Cl
IUPAC Name (Z)-1,4-dichlorobut-2-ene
Molecular Weight (g/mol) 124.992
PubChem CID 6432115
Formula Weight 125
Percent Purity ≥94%
Physical Form Liquid
Chemical Name or Material cis-1, 4-Dichloro-2-butene
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • cis-1,4-Dichloro-2-butene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1A
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H314-Causes severe skin burns and eye damage.
  • H350-May cause cancer.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.