missing translation for 'onlineSavingsMsg'
Learn More

cis-4-Cyanocyclohexylamine hydrochloride, 97%, Thermo Scientific™

Product Code. 15458828
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15458828 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15458828 Supplier Thermo Scientific Alfa Aesar Supplier No. H62292.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
cis-4-Cyanocyclohexylamine hydrochloride is used in chemical research.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1387445-51-3
Molecular Formula C7H13ClN2
Molecular Weight (g/mol) 160.645
MDL Number MFCD22378474
InChI Key BTLVSLNZSMIPCW-UHFFFAOYSA-N
Synonym trans-4-aminocyclohexanecarbonitrile hydrochloride, cis-4-aminocyclohexanecarbonitrile hydrochloride, 4-amino-cyclohexanecarbonitrile hydrochloride, 4-aminocyclohexanecarbonitrile hydrochloride, 4-aminocyclohexane-1-carbonitrile hydrochloride, c7h12n2.hcl, 1s,4s-4-aminocyclohexanecarbonitrile hydrochloride, 1r,4r-4-aminocyclohexane-1-carbonitrile hydrochloride, 1s,4s-4-aminocyclohexane-1-carbonitrile hydrochloride, cyclohexanecarbonitrile, 4-amino-, monohydrochloride, trans
PubChem CID 53407876
IUPAC Name 4-aminocyclohexane-1-carbonitrile;hydrochloride
SMILES C1CC(CCC1C#N)N.Cl

Specifications

Quantity 250 mg
UN Number UN3439
Formula Weight 160.64
Percent Purity 97%
Chemical Name or Material cis-4-Cyanocyclohexylamine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.