missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Citidine Deaminase Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-31412
This item is not returnable.
View return policy
Description
Citidine Deaminase Polyclonal specifically detects Citidine Deaminase in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Citidine Deaminase | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7), 20% Glycerol with 0.025% Proclin 300 | |
| CDDCytidine aminohydrolase, cytidine deaminase, cytosine nucleoside deaminase, EC 3.5.4.5, small cytidine deaminase | |
| Rabbit | |
| 16 kDa | |
| 100 μL | |
| Lipid and Metabolism | |
| 978 | |
| Human, Mouse | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1.0mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| P32320 | |
| CDA | |
| Recombinant protein encompassing a sequence within the center region of human CDA. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human. Predicted cross-reactivity based on sequence identity: Mouse (86%), Rat (86%), Bovine (86%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction