missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ CLA IdentiFiler™ Direct PCR Amplification Kit

Product Code. 31722920 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
200 x 25 μL Reactions
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
31722920 200 x 25 μL Reactions 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31722920 Supplier Applied Biosystems™ Supplier No. A65908
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

The CLA (Cell Line Authentication) IdentiFiler Direct PCR Amplification Kit simplifies the amplification step during authentication of cell lines.

The CLA (Cell Line Authentication) IdentiFiler Direct PCR Amplification Kit simplifies the amplification step during authentication of cell lines. The kit enables direct amplification of the same 16 loci included in our AmpFLSTR Identifiler Direct kit, eliminating the need to perform DNA extraction or purification steps for DNA database and other single-source reference samples. Blood or buccal samples on FTA cards can be punched into PCR plates or tubes, and taken directly to PCR amplification, without reduction in data quality. Samples collected using a Buccal DNA Collector or other non-FTA substrate can be processed without heat incubation or additional workflow steps using Prep-n-Go Buffer (available separately). The CLA Identifiler kit includes GeneScan 600 LIZ Size Standard v2.0, a five-dye-labeled, high-density size standard for the reproducible sizing of fragment analysis data.

Features of the CLA IdentiFiler Direct PCR Amplification Kit include:
• 5-dye, 16-locus STR analysis kit compatible with all cell line databasing loci standards
• Improved version of legacy AmpFLSTR Identifiler Direct kit
• Size standard included for ease of purchase
• Compatible with SeqStudio, 3730/xL, and 3500/xL genetic analyzers

Cell line authentication using short tandem repeat (STR) analysis kits can be used both for human sample authentication (HSA) and mixed sample analysis (MSA). In HSA, there is a need to verify the DNA profile of the sample to check for sample cross-contamination or sample mix-up, or to perform a quality check. For MSA there is a need to deconvolute the presence of multiple contributors in a sample (chimerism) through identification of multiple DNA genotypes in the sample.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • IdentiFiler Direct PCR Amplification Kit
• Liz600 Size Standard

Detection Method Gel Electrophoresis
Form Liquid
Format Kit
GC-Rich PCR Performance High
PCR Method Direct PCR
Polymerase AmpliTaq Gold™ DNA Polymerase
Reaction Speed 60 min.
Technique STR (Short Tandem Repeats) Analysis
For Use With (Equipment) SeqStudio Flex™ 3500, SeqStudio™ 3730xl Genetic Analyzers
Kit Type Multicomponent
Label or Dye LIZ, FAM, NED, PET, VIC
Quantity 200 x 25 μL Reactions
Species Human
Target Organism Class Cell lines
For Use With (Application) Genotyping, Cell Analysis
Isolation Technology Direct PCR
No. of Reactions 200 x 25 μL Reactions
Purification Target Genomic DNA from human cells
Reaction Format Master Mix
Sample Type Cell Line Culture
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.