Learn More
DWK Life Sciences™ Clamp for Spherical Joints, with tightening screw, made of stainless steel
Product Code. 10409342 Shop All DWK Life Sciences Products
Click to view available options
Size:
S 13
S 19
S 29
S 35
S 40 and S 41
S 51
S 64
Unit Size:
Each
Pack of 10
Pack of 25
Pack of 5
Pack of 50
Product Code. 10409342
Brand: DWK Life Sciences™ 286333806
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Ball joint clamp with additional locking option.
The ball joint clamp with locking screw enables a very secure connection by additional locking with the locking screw.
  • The locking screw enables a very secure connection by means of an additional locking device.
  • Excellent dimensional stability compared to conventional wire clamps
TRUSTED_SUSTAINABILITY
Specifications
Quantity 50
Material Stainless Steel
For Use With (Equipment) Securing of Spherical Ground Joints
For Use With (Application) Securing of spherical ground joints
Size S 29