Learn More
Corning™ Clamps for Erlenmeyer Flask
Product Code. 11313815 Shop All Corning Products
Change view
Click to view available options
Holds:
25 mL Erlenmeyer Flask
50 mL Erlenmeyer Flask
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds Unit Size
11313815 50 mL Erlenmeyer Flask Each
11303815 25 mL Erlenmeyer Flask Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11313815 Supplier Corning™ Supplier No. 480111
  • ONLINE EXCLUSIVE PRICE
    51.40€
    48.00€ / Pack of 1
    Save 3.40€
    6% Off
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
For use with various sizes of Erlenmeyer flasks.
This is a clamp for use with Erlenmeyer flasks, for use with Corning™ LSE™ Orbital Shaker and Corning™ LSE™ Low Speed Orbital Shaker.
TRUSTED_SUSTAINABILITY
Specifications
Holds 50 mL Erlenmeyer Flask
Product Type Clamp