Learn More
Claspin Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 23206-1-AP-20UL
 View more versions of this product
Product Code. 16802245
  • 145.00€ / 20µL
Description
Xenopus claspin is an essential upstream regulator of checkpoint kinase 1 and triggers a checkpoint arrest of the cell cycle in the presence of DNA templates in Xenopus egg extracts. The human gene appears to be the homolog Xenopus claspin and its function has not been determined.
Specifications
Specifications
Claspin
Polyclonal
Unconjugated
CLSPN
CLASPIN, CLSPN, hClaspin
Rabbit
Antigen Affinity Chromatography
RUO
63967
-20°C
Liquid
Immunoprecipitation, Immunocytochemistry, Western Blot, Immunofluorescence
0.29 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9HAW4
CLSPN
Claspin Fusion Protein Ag19637