Learn More
Fisherbrand Class II Biological Safety Cabinet
Product Code. 30564669 Shop All Fisherbrand Electrochemistry Products
Change view
Click to view available options
Length (Metric) Exterior:
1.2 m
1.8 m
Plug Type:
CEE 7/7
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Exterior Plug Type Unit Size
30564669 1.2 m CEE 7/7 Each
30564664 1.8 m CEE 7/7 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: This product is under eco-contribution tax.
Product Code. 30564669
Support a Safer, More Efficient Laboratory
Use promo code 28026
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: This product is under eco-contribution tax.
Fisherbrand Class II Biological Safety Cabinet is an economical solution for high protection.
  • Fisherbrand Class II biological safety cabinet (BSC) provides both personal and product protection from biological hazards and contaminations.
  • This model is available in 2 sizes; 1.2 m and 1.8 m with 3 different plug and socket options (Type F, Type G, and Type E)
  • Equipped with technology to automate airflow compensation and alerts user to out of tolerance airflows that could compromise product and operator protections
  • H14 HEPA filters removes airborne contaminants
  • 10-degrees sloped front for exceptional comfort and ergonomics
  • SmartClean front window design to improve access to the work area for cleaning
  • Night Set Back Mode reduces airflow during low-usage periods
  • The BSC's comply with all applicable performance and safety requirements of EN12469:2000
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN 12469
Chamber Material Stainless Steel (304)
Product Type Class II Biosafety Cabinet
Model Fisherbrand BSC 1.2
Noise Level <59 dBA
Outlets 2
Plug Type CEE 7/7
Power Consumption 1440 W
Receptacles 2 Single, Rear Wall
Exhaust Air Filter HEPA H14 EN1822
Filter Efficiency 99.995% at MPPS
Includes UV light, armrest and adjustable floor stand
Energy Consumption (Reduced Flow Mode) 70 W
Lighting LED
Ports 2 x 2
Supply Air Filter HEPA H14 EN1822
Warranty 2 Years
Worksurface Area 120 x 45 cm
Dimensions (L x W x H) Exterior 130 x 80.2 x 152 cm
Length (Metric) Exterior 1.2 m
Depth (Metric) Interior 63 cm
Electrical Requirements 230 V, 50/60 Hz
Voltage 230 V
Energy Consumption (Operating Set Point) 200 W
Quantity 1 Ea.