Learn More
Abeba™ Classic 1000 Shoes
Animal fats, oil and fuel resistant. Leather/microfiber construction.
Brand:  Abeba™ 1000/45
Additional Details : Weight : 0.95000kg
 View more versions of this product
Product Code. 15636567
  • 80.64€ / 1 pair
Estimated Shipment: 31-05-2024
to see stock.
Add to basket
Description
Description
  • CE EN ISO 20345:2011 SB A, E, FO SRC
  • Grained leather
  • White
  • Steel toe cap
  • Slip resistance PU outsole sole
  • Sanitized treated
  • Antistatic
  • Heel strap non-foldable, adjustable
  • Lining microfiber, respiration absorbing
  • Adjustable instep area
  • Buckle fastening
  • Heel patch microfiber
Specifications
Specifications
CE EN ISO 20345:2011 SB A, E, FO SRC
Microfiber
45
Polyurethane
Steel
White
Leather
Slip-resistant
1000
Animal fats, oil and fuel