Learn More
Veolia Water Systems™ Classic Polisher
Product Code. 11904937
Change view
Click to view available options
Depth (Metric):
37 cm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Depth (Metric) Unit Size
11904937 37 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Product Code. 11904937 Supplier Veolia Water Systems™ Supplier No. CLXXXDIM2
  • ONLINE EXCLUSIVE PRICE
    6273.75€
    4940.04€ / Pack of 1
    Save 1333.71€
    21% Off
 Restricted Item
Estimated Shipment: 23-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Specifications
Flow Rate 1.5 L/min.
Type Classic Polisher
Width (Metric) 41 cm
Height (Metric) 49 cm
Depth (Metric) 37 cm
Weight (Metric) 15 kg