Learn More
Kolmi Hopen™ Classic Polypropylene Hygiene Gown with Zipper
Product Code. 17790119 Shop All Medicom Products
Change view
Click to view available options
Size:
Large
2X-Large
Unit Size:
Pack of 50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17790119 2X-Large Pack of 50
17780119 Large Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17790119 Supplier Kolmi Hopen™ Supplier No. 40.635/05
  • ONLINE EXCLUSIVE PRICE
    71.36€
    60.34€ / Pack of 50
    Save 11.02€
    15% Off
Estimated Shipment: 14-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
These single-use gowns protect the working environment from particles coming from the operator, such as hair or sweat.
  • This Zipper-Closure Hygiene Gown protects the operator from work-related contamination
  • Also protects against sensitive products and processes that could be contaminated in a protected atmosphere
  • Pleasant and light, polypropylene fabric is breathable
  • Individually packaged for controlled hygiene and distribution
  • Without pockets to prevent falling objects
Specifications
Product Type General Purpose Gown
Size 2X-Large
Certifications/Compliance (Europe) ISO 9001
Gender Unisex
Material Polypropylene
Color Blue
Closure Type Zipper
Cuff Style Elastic
Disposable Single-use, Reusable
Sterility Non-sterile
Resistance Radioactive Particles, Infectious Agents
No. of Pockets None
Seam Type Sewn
Certifications/Compliance ISO 9001
Weight (Metric) 40 gsm
For Use With (Application) Category 1 Personal Protective Equipment
Packaging Type Individual, Bag
Autoclavable Not Autoclavable
Construction Made in China