Learn More
Cytiva Classical Powdered Ham's Nutrient Mixture F10 - F12
Description
Support cell culture growth and biomanufacturing processes with powdered HyClone™ Ham's F10-F12 Nutrient Mixture, manufactured using ISO 9001:2000-certified processes.
Specifications
Specifications
| Form | Powder |
| Product Type | Ham's F12, with L-glutamine |
| Sterilization Method | Non-sterile |
| With Additives | Glutamine |
| Without Additives | No Sodium Bicarbonate |
| Quantity | 1 x 50 L |
| Certifications/Compliance | cGMP and ISO 9001:2000 |
| Content And Storage | 2° to 8°C, away from light |
| Description | Ham's F12, with L-Glutamine |