missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Claudin-1 Antibody (CL3698) [Alexa Fluor™ 700], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61630AF700
This item is not returnable.
View return policy
Description
Claudin-1 Monoclonal antibody specifically detects Claudin-1 in Human samples. It is validated for Western Blot
Specifications
| Claudin-1 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| claudin 1, claudin-1, CLD1, ILVASCSenescence-associated epithelial membrane protein, SEMP1senescence-associated epithelial membrane protein 1 | |
| This antibody was developed using a recombinant protein derived from O95832, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Extracellular Matrix | |
| 9076 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Western Blot | |
| CL3698 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction