missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Claudin-11 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
378.00€ - 573.00€
Specifications
| Antigen | Claudin-11 |
|---|---|
| Dilution | Western Blot 0.04 - 0.4 ug/mL, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18483101
|
Novus Biologicals
NBP1-82470-25ul |
25 μL |
378.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18008605
|
Novus Biologicals
NBP1-82470 |
0.1 mL |
573.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Claudin-11 Polyclonal specifically detects Claudin-11 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Claudin-11 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2), 40% Glycerol with 0.02% Sodium Azide | |
| 5010 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:DAQAFGENRFYYTAGSSSPTHAKSAHV | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot 0.04 - 0.4 ug/mL, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Rabbit | |
| Human, Mouse | |
| claudin 11, claudin-11, Oligodendrocyte-specific protein, OSPOTMoligodendrocyte transmembrane protein | |
| CLDN11 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title