missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Claudin 23 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25296-1-AP-150UL
This item is not returnable.
View return policy
Description
CLDN23 is a four-transmembrane protein with WWCC motif, defined by W-X(17-22)-W-X(2)-C-X(8-10)-C. It is a candidate tumor suppressor gene implicated in intestinal-type gastric cancer.Specifications
| Claudin 23 | |
| Polyclonal | |
| Unconjugated | |
| CLDN23 | |
| claudin 23, CLDN23, CLDNL, hCG1646163 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 137075 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.37 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q96B33 | |
| CLDN23 | |
| Claudin 23 Fusion Protein Ag18189 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction