missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Claudin-3 Antibody (CL13128) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-18577JF669
This item is not returnable.
View return policy
Description
Claudin-3 Monoclonal antibody specifically detects Claudin-3 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| Claudin-3 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| C7orf1, claudin 3, claudin-3, Clostridium perfringens enterotoxin receptor 2, CPE-R 2, CPE-R2, CPETR2CPE-receptor 2, HRVP1, Rat ventral prostate.1 protein homolog, RVP1, ventral prostate.1 protein homolog, ventral prostate.1-like protein | |
| This antibody was developed using a synthetic peptide derived from O15551, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cell Biology, Cellular Markers, Extracellular Matrix | |
| 1365 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CL13128 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction