missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Claudin 3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16456-1-AP-150UL
This item is not returnable.
View return policy
Description
Claudin 1-8 proteins are a family of transmembrane proteins associated with tight junctions. Tight junctions are specialized regions of cell to cell contact; made up of network of strands to act as a molecular gasket for preventing the leakage of ions, water etc. between cells. They are abundant in luminal epithelial sheets where they maintain epithelial cell polarity. Different tissues exhibit different Claudin composition.Specifications
| Claudin 3 | |
| Polyclonal | |
| Unconjugated | |
| CLDN3 | |
| C7orf1, claudin 3, Claudin3, CLDN3, CPE R 2, CPE R2, CPE receptor 2, CPETR2, HRVP1, RVP1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 12739, 1365, 65130 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O15551, Q63400, Q9Z0G9 | |
| Cldn3 | |
| Claudin 3 Fusion Protein Ag9411 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction