missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Claudin-6 Antibody (9H8), Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals NBP3-26651-50ul
This item is not returnable.
View return policy
Description
Claudin-6 Monoclonal antibody specifically detects Claudin-6 in Human samples. It is validated for Flow Cytometry, ELISA
Specifications
| Claudin-6 | |
| Monoclonal | |
| Unconjugated | |
| 50% Glycerol, 0.01M PBS, PH 7.4 | |
| Human | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA | |
| 9H8 | |
| Flow Cytometry, ELISA | |
| claudin 6, claudin-6, skullin | |
| Recombinant Human Claudin-6 protein [UniProt P56747] | |
| 50 μL | |
| Stem Cell Markers | |
| 9074 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction