Learn More
Liofilchem™ Clean Test
Product Code. 11984359
Encompass_Preferred
Change view
Click to view available options
Detects:
Protein Fat And Sugar Residues
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Detects unitSize
11984359 Protein Fat And Sugar Residues Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11984359 Supplier Liofilchem™ Supplier No. 86001
Rapid test for the hygiene control of work surfaces
  • Clean test is a swab for the detection of protein, fat and sugar residues suitable for the check of work surfaces cleaning level.
Specifications
Detects Protein Fat And Sugar Residues
Manufacturing Origin Italy
Product Type Clean Test
Sterility Non-sterile
For Use With (Application) For The Hygiene Control Of Work Surfaces