Learn More
BINDER™ Cleaning Kit
Product Code. 16484019 Shop All BINDER Products
Change view
Click to view available options
Material:
Plastic
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material unitSize
16484019 Plastic Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16484019 Supplier BINDER™ Supplier No. 80120503
This Cleaning Kit is designed for maintenance and disinfection of BINDER™ equipment.
  • For equipment maintenance and disinfection
  • Consists of pH-neutral detergent (1000 g, concentrate) and spray disinfectant (500 mL, ready-to-use solution)
  • Also includes lint-free cleansing tissues, protective gloves, and protective glasses
Specifications
Description Cleaning kit - for equipment maintenance and disinfection, consisting of pH-neutral detergent (1000 g, concentrate), spray disinfectant (500 mL, ready-to-use solution), lint-free cleansing tissues, protective gloves, and protective glasses
For Use With (Equipment) BINDER Chambers Series CB-S
Material Plastic
Type Cleaning Kit
For Use With (Application) For Equipment Maintenance and Disinfection
Includes pH-neutral Detergent (1000 g, concentrate), Spray Disinfectant (500 mL, ready-to-use solution), Lint-free Cleansing Tissues, Protective Gloves, and Protective Glasses