Description
- For equipment maintenance and disinfection
- Consists of pH-neutral detergent (1000 g, concentrate) and spray disinfectant (500 mL, ready-to-use solution)
- Also includes lint-free cleansing tissues, protective gloves, and protective glasses
Specifications
Specifications
| Description | Cleaning kit - for equipment maintenance and disinfection, consisting of pH-neutral detergent (1000 g, concentrate), spray disinfectant (500 mL, ready-to-use solution), lint-free cleansing tissues, protective gloves, and protective glasses |
| For Use With (Equipment) | BINDER Chambers Series CB-S |
| Material | Plastic |
| Type | Cleaning Kit |
| For Use With (Application) | For Equipment Maintenance and Disinfection |
| Includes | pH-neutral Detergent (1000 g, concentrate), Spray Disinfectant (500 mL, ready-to-use solution), Lint-free Cleansing Tissues, Protective Gloves, and Protective Glasses |