Learn More
Cole-Parmer Cleanroom Hose Barb Fittings, Y Union, Animal-Derivative-Free Polypropylene
Product Code. 30053789 Shop All Cole-Parmer Products
Change view
Click to view available options
Diameter (English) Inner:
1 in.
1/16 in.
1/2 in.
1/4 in.
1/8 in.
3/16 in.
3/32 in.
3/4 in.
3/8 in.
5/16 in.
5/32 in.
5/8 in.
Unit Size:
Pack of 10
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (English) Inner unitSize
30053789 1/4 in. Pack of 10
30053392 3/32 in. Pack of 10
30053795 5/8 in. Pack of 10
30054203 1/8 in. Pack of 10
30053028 1 in. Pack of 10
30053656 3/4 in. Pack of 10
30052991 5/16 in. Pack of 10
30053253 1/2 in. Pack of 10
30054164 3/8 in. Pack of 10
30053243 5/32 in. Pack of 10
30053979 3/16 in. Pack of 10
30053599 1/16 in. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Product Code. 30053789 Supplier Cole-Parmer Supplier No. 5011142
Only null left
Add to basket
Add to basket
Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Quick and easy assembly without tools. Single barb design tightly grips the tubing. Meets FDA requirements 21 CFR 177.1520(a)(3)(i) and (c)3.1a and RoHS compliant
Specifications
Certifications/Compliance ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS
Diameter (English) Inner 1/4 in.
Min. Operating Temperature (Metric) -23°C
Packaging Quantity 10/Pk.
Outlet Port Hose Barb Y
Sterilization Method 121°C
Shape Y
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.