Learn More
Cole-Parmer Cleanroom Male Luer Lock Plug and Nut Assemblies, Straight Adapter, White Nylon
Product Code. 30053703 Shop All Cole-Parmer Products
Change view
Click to view available options
Color:
Berry
Lime
Natural
Orange
Rose
Violet
Yellow
Unit Size:
Pack of 10
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
30053703 Yellow Pack of 10
30052930 Violet Pack of 10
30054317 Berry Pack of 10
30054267 Natural Pack of 10
30054012 Orange Pack of 10
30053245 Lime Pack of 10
30053675 Rose Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 30053703 Supplier Cole-Parmer Supplier No. 5011048
Only null left
Add to basket
Add to basket
Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Nut and body rotate independently during connection
Specifications
Certifications/Compliance FDA;Non-DEHP;Non-Phthalate;RoHS
Material Nylon
Inlet Male Luer Lock, Plug
Color Yellow