Learn More
Cole-Parmer Cleanroom Male Luer Lock Plug and Nut Assemblies, Straight Adapter, White Nylon
Shop All Cole-Parmer ProductsDescription
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Nut and body rotate independently during connection
Specifications
Specifications
| Certifications/Compliance | FDA;Non-DEHP;Non-Phthalate;RoHS |
| Material | Nylon |
| Inlet | Male Luer Lock, Plug |
| Color | Yellow |