missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CLEC4G Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5148644
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
This gene encodes a glycan-binding receptor and member of the C-type lectin family which plays a role in the T-cell immune response. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
| CLEC4G | |
| Polyclonal | |
| Unconjugated | |
| CLEC4G | |
| CLEC4G; C-type lectin domain family 4 member G; C-type lectin domain family 4, member G; C-type lectin superfamily 4, member G; DTTR431; Liver and lymph node sinusoidal endothelial cell C-type lectin; LOC100066617; LP2698; LSECtin; UNQ431; UNQ431/PRO792 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 339390 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| Q6UXB4 | |
| CLEC4G | |
| Mouse myeloma cell line NS0-derived recombinant human LSECtin/CLEC4G, Ser54-Cys293. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction