missing translation for 'onlineSavingsMsg'
Learn More

Clencyclohexerol, TRC

Product Code. 30010262
Change view
Click to view available options
Quantity:
1 mg
10 mg
25 mg
Conditionnement:
10mg
1mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantity unitSize
30010262 10 mg 10mg
30004651 25 mg 25mg
30007668 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 30010262 Fournisseur Toronto Research Chemicals Code fournisseur TRCC57250010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 157877-79-7
Molecular Formula C14 H20 Cl2 N2 O2
Molecular Weight (g/mol) 319.23
Synonym 4-Amino-3,5-dichloro-α-[[(4-hydroxycyclohexyl)amino]methyl]benzenemethanol, Clencyclohexerol, Benzenemethanol, 4-amino-3,5-dichloro-α-[[(4-hydroxycyclohexyl)amino]methyl]-
IUPAC Name 4-[[2-(4-amino-3,5-dichlorophenyl)-2-hydroxyethyl]amino]cyclohexan-1-ol
SMILES Nc1c(Cl)cc(cc1Cl)C(O)CNC2CCC(O)CC2

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C14H20Cl2N2O2/c15-11-5-8(6-12(16)14(11)17)13(20)7-18-9-1-3-10(19)4-2-9/h5-6,9-10,13,18-20H,1-4,7,17H2
Formula Weight 318.0902
Percent Purity >95%
Chemical Name or Material Clencyclohexerol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.