missing translation for 'onlineSavingsMsg'
Learn More

Clenproperol, TRC

Product Code. 30004166
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30004166 250 mg 250mg
30012823 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30004166 Supplier Toronto Research Chemicals Supplier No. TRCC573000250MG
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38339-11-6
Molecular Formula C11 H16 Cl2 N2 O
Molecular Weight (g/mol) 263.16
Synonym 4-Amino-3,5-dichloro-α-[[(1-methylethyl)amino]methyl]benzenemethanol, 4-Amino-3,5-dichloro-α-[(isopropylamino)methyl]benzyl alcohol, Clenproperol, NAB 277
IUPAC Name 1-(4-amino-3,5-dichlorophenyl)-2-(propan-2-ylamino)ethanol
SMILES CC(C)NCC(O)c1cc(Cl)c(N)c(Cl)c1

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C11H16Cl2N2O/c1-6(2)15-5-10(16)7-3-8(12)11(14)9(13)4-7/h3-4,6,10,15-16H,5,14H2,1-2H3
Formula Weight 262.064
Percent Purity >95%
Chemical Name or Material Clenproperol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.