missing translation for 'onlineSavingsMsg'
Learn More

Cletoquine, TRC

Product Code. 17904605
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17904605 5 mg 5mg
17990775 1 mg 1mg
17921695 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17904605 Supplier Toronto Research Chemicals Supplier No. TRCC5735055MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4298-15-1
Molecular Formula C16 H22 Cl N3 O
Molecular Weight (g/mol) 307.82
Synonym Ethanol, 2-[[4-[(7-chloro-4-quinolinyl)amino]pentyl]amino]-, Ethanol, 2-[[4-[(7-chloro-4-quinolyl)amino]pentyl]amino]- (7CI,8CI), 2-[[4-[(7-Chloro-4-quinolinyl)amino]pentyl]amino]ethanol, (±)-Desethylhydroxychloroquine, Cletoquine, Desethylhydroxychloroquine
IUPAC Name 2-[4-[(7-chloroquinolin-4-yl)amino]pentylamino]ethanol
SMILES CC(CCCNCCO)Nc1ccnc2cc(Cl)ccc12

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C16H22ClN3O/c1-12(3-2-7-18-9-10-21)20-15-6-8-19-16-11-13(17)4-5-14(15)16/h4-6,8,11-12,18,21H,2-3,7,9-10H2,1H3,(H,19,20)
Formula Weight 307.1451
Percent Purity >95%
Chemical Name or Material Cletoquine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.