Learn More
Clidinium Bromide, Mikromol™
Description
Clidinium Bromide
Specifications
Specifications
| Chemical Name or Material | Clidinium Bromide |
| CAS | 3485-62-9 |
| Analyte or Component Names | Clidinium Bromide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C22 H26 N O3 . Br |
| Certifications/Compliance | ISO 17025 |
| Synonym | Clidinium Bromide, 1-Azoniabicyclo[2.2.2]octane, 3-[(2-hydroxy-2,2-diphenylacetyl)oxy]-1-methyl-, bromide (1:1), 1-Azoniabicyclo[2.2.2]octane, 3-[(hydroxydiphenylacetyl)oxy]-1-methyl-, bromide (9CI), 3-Hydroxy-1-methylquinuclidinium bromide, benzilate (6CI,7CI), Quinuclidinium, 3-hydroxy-1-methyl-, bromide, benzilate (8CI), 1-Methyl-3-(benziloyloxy)quinuclidinium bromide, 3-(Benziloyloxy)-1-methylquinuclidinium bromide, Clidinium bromide, Quarzan, Quarzan bromide, Ro 2-3773 |
| InChI Formula | InChI=1S/C22H26NO3.BrH/c1-23-14-12-17(13-15-23)20(16-23)26-21(24)22(25,18-8-4-2-5-9-18)19-10-6-3-7-11-19;/h2-11,17,20,25H,12-16H2,1H3;1H/q+1;/p-1 |
| SMILES | [Br-].C[N+]12CCC(CC1)C(C2)OC(=O)C(O)(c3ccccc3)c4ccccc4 |
| IUPAC Name | (1-methyl-1-azoniabicyclo[2.2.2]octan-3-yl) 2-hydroxy-2,2-diphenylacetate;bromide |
| Show More |