Learn More
CLINT1 Monoclonal Antibody (OTI9E1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA805981
Description
Binds to membranes enriched in phosphatidylinositol 4,5- bisphosphate (PtdIns(4,5)P2). May have a role in transport via clathrin-coated vesicles from the trans-Golgi network to endosomes. Stimulates clathrin assembly.Specifications
| CLINT1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q14677 | |
| CLINT1 | |
| Human recombit protein fragment corresponding to amino acids 1-290 of human CLINT1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI9E1 | |
| Unconjugated | |
| CLINT1 | |
| CLINT, ENTH, EPN4, EPNR | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 9685 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.