missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLIP Mouse anti-Human, BB700, Clone: CerCLIP, BD OptiBuild™
Mouse Monoclonal Antibody
Brand: BD Biosciences 745964
Additional Details : Weight : 0.01000kg
Description
CLIP Monoclonal antibody specifically detects CLIP in Human samples. It is validated for Flow Cytometry.Specifications
CLIP | |
Monoclonal | |
0.2 mg/mL | |
Mouse | |
RUO | |
Human | |
Purified |
Flow Cytometry | |
CerCLIP | |
Brilliant Blue 700 (BB700) | |
50 μg | |
Primary | |
Refrigerate (2 to 8°C) | |
IgG1 κ |