missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLIP Mouse anti-Human, BUV805, Clone: CerCLIP, BD Biosciences™
Mouse Monoclonal Antibody
Brand: BD Biosciences 748549
Additional Details : Weight : 0.00030kg
Description
CLIP Monoclonal antibody specifically detects CLIP in Human samples. It is validated for Flow CytometrySpecifications
CLIP | |
Monoclonal | |
Brilliant Ultraviolet 805 | |
Mouse | |
RUO | |
Human | |
IgG1 κ |
Flow Cytometry | |
CerCLIP | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV805 under optimal conditions that minimize unconjugated dye and antibody. |