Learn More
Gilac™ Clip-on Lid for Dough Container
Product Code. 17988541
Change view
Click to view available options
For Use With (Equipment):
Dough Container
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
17988541 Dough Container Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17988541 Supplier Gilac™ Supplier No. G179211
  • ONLINE EXCLUSIVE PRICE
    13.64€
    8.99€ / Pack of 1
    Save 4.65€
    34% Off
Estimated Shipment: 26-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Clip-on Lid for Dough Container is a lid for half size dough container.
  • Lid for half size dough container.
Specifications
Product Type Lid
Material Polyethylene
Color White
Dimensions (L x W x H) 400 x 300 x 30 mm
For Use With (Equipment) Dough Container