missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Clofarabine
Description
Clofarabine is a deoxycytidine kinase (dCK) substrate. Phosphorylated to form clofarabine triphosphate, which competes with dATP for DNA polymerase-α and -ε and potently inhibits ribonucleotide reductase (IC50 = 65 nM). Induces apoptosis by directly altering mitochondrial transmembrane potential. Demonstrates growth inhibition and cytotoxic activity in a variety of leukemias and solid tumors.
Specifications
Specifications
| Chemical Name or Material | 2-Chloro-9-(2-deoxy-2-fluoro-β-D-arabinofuranosyl)-9H-purin-6-amine |
| CAS | 123318-82-1 |
| Assay Percent Range | >99% |
| Quantity | 5 mg |
| Product Line | Tocris |
| Molecular Formula | C10H11ClFN5O3 |
| Formula Weight | Observed MW: 303.68 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?