missing translation for 'onlineSavingsMsg'
Learn More

Cloperastine Fendizoate, TRC

Product Code. 30011896
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011896 100 mg 100mg
30008700 500 mg 500mg
30007567 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011896 Supplier Toronto Research Chemicals Supplier No. TRCC584455100MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85187-37-7
Molecular Formula C20 H24 Cl N O . C20 H14 O4
Molecular Weight (g/mol) 648.19
Synonym 1-[2-[(4-Chlorophenyl)phenylmethoxy]ethyl]-hydroxy[1,1'-biphenyl]-3-yl)carbonyl]benzoate Piperidine
IUPAC Name 1-[2-[(4-chlorophenyl)-phenyl-methoxy]ethyl]piperidine;2-(4-hydroxy-3-phenyl-benzoyl)benzoic acid
SMILES OC(=O)c1ccccc1C(=O)c2ccc(O)c(c2)c3ccccc3.Clc4ccc(cc4)C(OCCN5CCCCC5)c6ccccc6

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C20H24ClNO.C20H14O4/c21-19-11-9-18(10-12-19)20(17-7-3-1-4-8-17)23-16-15-22-13-5-2-6-14-22;21-18-11-10-14(12-17(18)13-6-2-1-3-7-13)19(22)15-8-4-5-9-16(15)20(23)24/h1,3-4,7-12,20H,2,5-6,13-16H2;1-12,21H,(H,23,24)
Formula Weight 647.2439
Percent Purity >95%
Chemical Name or Material Cloperastine Fendizoate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.