missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Clostridium perfringens Perfringolysin O Polyclonal Antibody
Shop All Thermo Scientific Products

Description
This antibody has been tested in direct-ELISA.
Sulfhydryl-activated toxin that causes cytolysis by forming pores in cholesterol containing host membranes. After binding to target membranes, the protein assembles into a pre-pore complex. A conformation change leads to insertion in the host membrane and formation of an oligomeric pore complex. Cholesterol may be required for binding to host cell membranes, membrane insertion and pore formation. Can be reversibly inactivated by oxidation.
Specifications
Specifications
| Antigen | Clostridium perfringens Perfringolysin O |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 |
| Gene Alias | pfo; pfoA; pfoR; Theta-toxin; Thiol-activated cytolysin |
| Host Species | Rabbit |
| Immunogen | Recombinant Clostridium perfringens Perfringolysin O protein (29-500aa). |
| Purification Method | Protein G |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?