Learn More
Lenz Laborglasintrumente™ Closure for Stirrer
Product Code. 10285223
Change view
Click to view available options
Cone Size:
NS 29/32
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cone Size Unit Size
10285223 NS 29/32 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10285223 Supplier Lenz Laborglasintrumente™ Supplier No. 06310504
  • ONLINE EXCLUSIVE PRICE
    92.00€
    59.95€ / Pack of 1
    Save 32.05€
    34% Off
Estimated Shipment: 24-09-2026
Add to basket
Add to basket
Closure for shaft with blade.
  • Spare closure only
Specifications
Cone Size NS 29/32
Description Closure
For Use With (Equipment) For round bottom flasks 250 to 2000 ml
Type Thread GL 25
Material Duran