Learn More
CNKSR1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA556574
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: SPRTSFGSLT DSSEEALEGM VRGLRQGGVS LLGQPQPLTQ EQWRSSFMRR NRDPQLNERV HRVRALQSTL KAKLQELQVL EEVLGDPELT GEKFRQWKEQ NRE Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 90%.
This gene encodes a protein containing several motifs involved in protein-protein interaction, including PDZ, PH (Pleckstrin homology), and SAM (sterile alpha motif) domains. The encoded protein acts as a scaffold component for receptor tyrosine kinase signaling and may mediate crosstalk between different signaling pathways. Alternative splicing results in multiple transcript variants.Specifications
CNKSR1 | |
Polyclonal | |
Unconjugated | |
CNKSR1 | |
BC037758; CNK; CNK homolog protein 1; CNK1; CNKSR1; Connector enhancer of kinase suppressor of ras 1; connector enhancer of KSR 1; Connector enhancer of KSR-like | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10256 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q969H4 | |
CNKSR1 | |
Recombinant protein corresponding to Human CNKSR1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only