missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CNKSR1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA5101230
Additional Details : Weight : 0.01000kg
Description
Antibody detects endogenous levels of total CNKSR1.
This gene encodes a protein containing several motifs involved in protein-protein interaction, including PDZ, PH (Pleckstrin homology), and SAM (sterile alpha motif) domains. The encoded protein acts as a scaffold component for receptor tyrosine kinase signaling and may mediate crosstalk between different signaling pathways. Alternative splicing results in multiple transcript variants.Specifications
CNKSR1 | |
Polyclonal | |
Unconjugated | |
CNKSR1 | |
BC037758; CNK; CNK homolog protein 1; CNK1; CNKSR1; Connector enhancer of kinase suppressor of ras 1; connector enhancer of KSR 1; Connector enhancer of KSR-like | |
Rabbit | |
Affinity chromatography | |
RUO | |
10256, 194231, 298545 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q969H4 | |
CNKSR1 | |
A synthesized peptide derived from human CNKSR1(Accession Q969H4), corresponding to amino acid residues L350-E400. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |