Learn More
CNKSR3 Monoclonal Antibody (OTI1D7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA504737
Description
CNKSR3 may be involved in transepithelial sodium transport. Regulates aldosterone-induced and ENaC-mediated sodium transport possibly through regulation of the ERK pathway.Specifications
| CNKSR3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q6P9H4 | |
| CNKSR3 | |
| Full length human recombit protein of human CNKSR3 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI1D7 | |
| Unconjugated | |
| CNKSR3 | |
| CNK3, CNK3/IPCEF1, MAGI1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 154043 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.