Learn More
CNKSR3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA562023
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: FLDQESRRRR FTIADSDQLP GYSVETNILP TKMREKTPSY GKPRPLSMPA DGNWMGIVDP FARPRGHGRK GEDALCRYFS NERIP Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 92%.
CNKSR3 may be involved in transepithelial sodium transport. Regulates aldosterone-induced and ENaC-mediated sodium transport possibly through regulation of the ERK pathway.Specifications
CNKSR3 | |
Polyclonal | |
Unconjugated | |
CNKSR3 | |
6820402C05; BC024086; CNK homolog protein 3; CNK3; CNKSR family member 3; Cnksr3; Connector enhancer of kinase suppressor of ras 3; connector enhancer of KSR 3; KOX14; MAGI1; maguin-like protein; membrane associated guanylate kinase interacting protein-like 1; membrane associated guanylate kinase, WW and PDZ domain containing 1; membrane-associated guanylate kinase-interacting protein-like 1; parturition-related protein 4; Prp4; ZNF21 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
154043 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6P9H4 | |
CNKSR3 | |
Recombinant protein corresponding to Human CNKSR3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only