missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CNPase Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
159.00€ - 464.00€
Specifications
| Antigen | CNPase |
|---|---|
| Dilution | Western Blot 1:1000 - 1:5000, Immunohistochemistry 1:1000 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:1000 - 1:2000 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18667908
|
Novus Biologicals
NBP2-50058SS |
0.025 mL |
159.00€
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18652268
|
Novus Biologicals
NBP2-50058 |
0.1 mL |
464.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CNPase Polyclonal antibody specifically detects CNPase in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ ImmunofluorescenceSpecifications
| CNPase | |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cellular Markers, Glia Markers, Neuronal Cell Markers, Neuroscience, Oligodendrocyte Cell Markers | |
| Supplied as serum | |
| 2', 3' cyclic nucleotide 3' phosphohydrolase, 2'-3'-cyclic nucleotide 3' phosphodiesterase, 2'-3'-cyclic-nucleotide 3'-phosphodiesterase, CNP1, CNPase, EC 3.1.4.37 | |
| Full length human CNPase recombinant protein. [UniProt# P09543] | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot 1:1000 - 1:5000, Immunohistochemistry 1:1000 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:1000 - 1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| P09543 | |
| 1267 | |
| IgG | |
| Immunogen affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title