missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CNTD1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24854-1-AP-20UL
This item is not returnable.
View return policy
Description
CNTD1 is a protein coding gene.Specifications
| CNTD1 | |
| Polyclonal | |
| Unconjugated | |
| CNTD1 | |
| CNTD, CNTD1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 124817, 68107 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8N815, Q9D995 | |
| Cntd1 | |
| CNTD1 Fusion Protein Ag21338 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction