Learn More
Aalborg™ CO2 Direct Flow Tube
Product Code. 11882450
Change view
Click to view available options
Airflow Rate:
1 L/min.
100 mL/min.
300 mL/min.
6 L/min.
Width (Metric):
65 mm
150 mm
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Airflow Rate Width (Metric) Unit Size
11882450 300 mL/min. 150 mm Each
11882270 6 L/min. 65 mm Each
11862250 1 L/min. 65 mm Each
11842450 100 mL/min. 150 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11882450 Supplier Aalborg™ Supplier No. FT06214STVN
  • ONLINE EXCLUSIVE PRICE
    123.90€
    75.48€ / Pack of 1
    Save 48.42€
    39% Off
 Restricted Item
Estimated Shipment: 27-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Aalborg™ CO2 Direct Flow Tube completes a flowmeter, which is used in fluid systems to indicate the rate of flow of the fluid.
  • Direct reading tubes are straightforward.
  • The measurement scale on each of these tubes reads actual flow at standard conditions in a choice of English or Metric units.
  • Obtaining actual flow rates with these tubes requires the use of a reference scale flow correlation table.
  • Reference scale tubes are useful when measuring flow rates for gases other than air and/or for non-standard conditions.
Specifications
Type CO2 Direct Flow Tube
Width (Metric) 150 mm
Airflow Rate 300 mL/min.