Learn More
Thermo Scientific™ CO2 and Other Incubator Calibration, ISO 9001
Product Code. 32018036 Shop All Thermo Scientific Products
Change view
Click to view available options
Duration:
1 Day
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Duration Unit Size
32018036 1 Day Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 32018036 Supplier Thermo Scientific™ Supplier No. CALBCINC
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
CO2 and other incubator calibration includes CO2, temperature and humidity. ISO 9001: Certificate with before and after data. Regulatory requirements: Ideal for verifying equipment performance to manufacturer’s specifications.
CO2 and other incubator calibration includes CO2, temperature and humidity
  • ISO 9001: Certificate with before and after data.
  • Regulatory requirements: Ideal for organization needing to verify equipment performance to manufacturer's specifications. Uncertainties not required.
  • Included in service delivery: Found as left calibration data; detailed calibration label, If a unit is out of tolerance an adjustment is performed
  • Included on accredited certificate: Serial/asset number; Measurement data; Environmental conditions; Conformity assessment; Reference equipment used
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) CO2 Incubators
Duration 1 Day
Product Type Calibration