Learn More
Coagulation Factor III/Tissue Factor Antibody (001), Janelia Fluor™ 646, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Coagulation Factor III/Tissue Factor Monoclonal antibody specifically detects Coagulation Factor III/Tissue Factor in Human samples. It is validated for ELISA
Specifications
Specifications
| Gene Alias | CD142, CD142 antigen, Coagulation factor III, coagulation factor III (thromboplastin, tissue factor), FLJ17960, TF, TFA, Thromboplastin, tissue factor |
| Content And Storage | Store at 4C in the dark. |