missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Coagulation Factor III/Tissue Factor Antibody (CL3805) - Azide and BSA Free, Novus Biologicals™
Shop All Bio Techne ProductsBeskrivelse
Coagulation Factor III/Tissue Factor Monoclonal antibody specifically detects Coagulation Factor III/Tissue Factor in Human samples. It is validated for Western Blot,Immunohistochemistry (Paraffin),Immunohistochemistry,KnockDown
Tekniske data
Tekniske data
| Antigen | Coagulation Factor III/Tissue Factor |
| Applications | Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry, KnockDown |
| Classification | Monoclonal |
| Clone | CL3805 |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1 μg/mL, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
| Formulation | Lyophilized in 5% Trehalose. |
| Gene Alias | CD142, CD142 antigen, Coagulation factor III, coagulation factor III (thromboplastin, tissue factor), FLJ17960, TF, TFA, Thromboplastin, tissue factor |
| Host Species | Mouse |
| Vis mere |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?