missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Coagulation Factor XIV/Protein C Antibody (PC 7), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-05113PCP
This item is not returnable.
View return policy
Description
Coagulation Factor XIV/Protein C Monoclonal antibody specifically detects Coagulation Factor XIV/Protein C in Human samples. It is validated for ELISA
Specifications
| Coagulation Factor XIV/Protein C | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| PC 7 | |
| ELISA | |
| Anticoagulant protein C, APC, Autoprothrombin IIA, Blood coagulation factor XIV, EC 3.4.21, EC 3.4.21.69, PC, PROC1, protein C (inactivator of coagulation factors Va and VIIIa), vitamin K-dependent protein C | |
| Recombinant human activated protein C | |
| 0.1 mL | |
| Apoptosis, Cancer | |
| 5624 | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction